C12H316CEEDE Chemcia Scientific, LLC

1-Chloro-3-iodo-5-methyl-benzene

  • MDL number MFCD13185456
  • Cas Number : 116632-43-0
  • Molecular Weight : 252.48327

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Red Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5295

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5295
Availability details for
Quantity
  • Availability
  • Add to Cart