C12H316CEEDF Chemcia Scientific, LLC

2-Amino-4,5-difluoro-benzaldehyde

  • MDL number MFCD18824350
  • Cas Number : 172843-46-8
  • Molecular Weight : 157.1208

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5365

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5365
Availability details for
Quantity
  • Availability
  • Add to Cart