C12H316CEEE2 Chemcia Scientific, LLC

5-Chloro-2-chloromethyl-6-nitro-benzooxazole

  • MDL number MFCD11844814
  • Cas Number : 1126637-40-8
  • Molecular Weight : 247.03868

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5370

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5370
Availability details for
Quantity
  • Availability
  • Add to Cart