C12H316CEEE3 Chemcia Scientific, LLC

5-Fluoro-4-methyl-2-nitro-phenol

  • MDL number MFCD16658619
  • Cas Number : 83341-28-0
  • Molecular Weight : 171.12917

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5371

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5371
Availability details for
Quantity
  • Availability
  • Add to Cart