C12H316CEEE4 Chemcia Scientific, LLC

5-Chloro-2-hydroxy-6-methyl-nicotinic acid

  • MDL number MFCD11044289
  • Cas Number : 117449-75-9
  • Molecular Weight : 187.58377

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5373

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5373
Availability details for
Quantity
  • Availability
  • Add to Cart