C12H316CEEE9 Chemcia Scientific, LLC

2-Chloro-4-methoxy-5-nitro-benzoic acid

  • MDL number MFCD26395064
  • Cas Number : 33458-99-0
  • Molecular Weight : 231.59372

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5319

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5319
Availability details for
Quantity
  • Availability
  • Add to Cart