C12H316CEEEC Chemcia Scientific, LLC

3-Nitro-imidazo[1,2-a]pyridine-5-carboxylic acid methyl ester

  • Cas Number : 88061-85-2
  • Molecular Weight : 221.17384

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5326

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5326
Availability details for
Quantity
  • Availability
  • Add to Cart