C12H316CEEED Chemcia Scientific, LLC

3-Amino-imidazo[1,2-a]pyridine-5-carboxylic acid methyl ester

  • Cas Number : 111753-15-2
  • Molecular Weight : 191.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-aminoimidazo[1,2-a]pyridine-5-carboxylate
Inchi Code 1S/C9H9N3O2/c1-14-9(13)6-3-2-4-8-11-5-7(10)12(6)8/h2-5H,10H2,1H3
Inchi Key YNSZOPQMEOIKRB-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Reddish Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5327

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5327
Availability details for
Quantity
  • Availability
  • Add to Cart