C12H316CEEFF Chemcia Scientific, LLC

2-Fluoro-4-hydroxy-5-nitro-benzonitrile

  • MDL number MFCD16658599
  • Cas Number : 1134198-38-1
  • Molecular Weight : 182.11196

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5361

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5361
Availability details for
Quantity
  • Availability
  • Add to Cart