C12H316CEF00 Chemcia Scientific, LLC

2-Fluoro-4-hydroxy-benzamide

  • MDL number MFCD09999525
  • Cas Number : 1133122-96-9
  • Molecular Weight : 155.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-fluoro-4-hydroxybenzamide
Inchi Code 1S/C7H6FNO2/c8-6-3-4(10)1-2-5(6)7(9)11/h1-3,10H,(H2,9,11)
Inchi Key ZOGOWOQZTFRRHO-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5369

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5369
Availability details for
Quantity
  • Availability
  • Add to Cart