C12H316CEF01 Chemcia Scientific, LLC

1,5-Difluoro-2-methyl-3-nitro-benzene

  • MDL number MFCD22557859
  • Cas Number : 1188412-98-7
  • Molecular Weight : 173.1202

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5378

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5378
Availability details for
Quantity
  • Availability
  • Add to Cart