C12H316CEF03 Chemcia Scientific, LLC

6-Methoxy-benzo[d]isoxazol-3-ylamine

  • MDL number MFCD18801052
  • Cas Number : 157368-82-6
  • Molecular Weight : 164.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-methoxy-1,2-benzisoxazol-3-ylamine
Inchi Code 1S/C8H8N2O2/c1-11-5-2-3-6-7(4-5)12-10-8(6)9/h2-4H,1H3,(H2,9,10)
Inchi Key UYWCFQVFVCXHCP-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5381

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5381
Availability details for
Quantity
  • Availability
  • Add to Cart