C12H316CEF04 Chemcia Scientific, LLC

6-Methoxy-1H-indazol-3-ylamine

  • MDL number MFCD07385621
  • Cas Number : 511225-17-5
  • Molecular Weight : 163.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-methoxy-1H-indazol-3-amine
Inchi Code 1S/C8H9N3O/c1-12-5-2-3-6-7(4-5)10-11-8(6)9/h2-4H,1H3,(H3,9,10,11)
Inchi Key HNOSKPXFWPKCPP-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5382

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5382
Availability details for
Quantity
  • Availability
  • Add to Cart