C12H316CEF05 Chemcia Scientific, LLC

6-Methoxy-2H-indazol-3-ylamine

  • MDL number MFCD28539849
  • Cas Number : 511225-17-5
  • Molecular Weight : 163.18043

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5383

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5383
Availability details for
Quantity
  • Availability
  • Add to Cart