C12H316CEF06 Chemcia Scientific, LLC

5-Amino-2-fluoro-4-hydroxy-benzonitrile

  • MDL number MFCD16658600
  • Cas Number : 388091-38-1
  • Molecular Weight : 152.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-amino-2-fluoro-4-hydroxybenzonitrile
Inchi Code 1S/C7H5FN2O/c8-5-2-7(11)6(10)1-4(5)3-9/h1-2,11H,10H2
Inchi Key HCWFDMJNYDNDOB-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5362

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5362
Availability details for
Quantity
  • Availability
  • Add to Cart