C12H316CEF07 Chemcia Scientific, LLC

Acetic acid 3-fluoro-4-methyl-phenyl ester

  • MDL number MFCD28968028
  • Cas Number : 1862507-17-2
  • Molecular Weight : 168.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-fluoro-4-methylphenyl acetate
Inchi Code 1S/C9H9FO2/c1-6-3-4-8(5-9(6)10)12-7(2)11/h3-5H,1-2H3
Inchi Key GXQCLFMLNUDFHF-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5363

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5363
Availability details for
Quantity
  • Availability
  • Add to Cart