C12H316CEF0B Chemcia Scientific, LLC

5-Amino-4-bromo-2-methoxy-benzoic acid methyl ester

  • Cas Number : 1548087-31-5
  • Molecular Weight : 260.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 5-amino-4-bromo-2-methoxybenzoate
Inchi Code 1S/C9H10BrNO3/c1-13-8-4-6(10)7(11)3-5(8)9(12)14-2/h3-4H,11H2,1-2H3
Inchi Key QQYPAFANRMGTHT-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5394

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5394
Availability details for
Quantity
  • Availability
  • Add to Cart