C12H316CEF10 Chemcia Scientific, LLC

6-Bromo-8-trifluoromethyl-imidazo[1,2-a]pyridine-2-carboxylic acid ethyl ester

  • MDL number MFCD197040~5C1
  • Cas Number : 1121051-30-6
  • Molecular Weight : 337.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5C
Shipping Temperature Ambient
Country of Origin USA
Product Link http://www.chemcia.com/product_fl02.asp?id=5400

Safety Information

msds http://www.chemcia.com/product_fl02.asp?id=5400
Availability details for
Quantity
  • Availability
  • Add to Cart