C12H316CEF12 Chemcia Scientific, LLC

2-tert-Butoxycarbonylamino-5-methoxy-benzoic acid

  • MDL number MFCD02682214
  • Cas Number : 262614-64-2
  • Molecular Weight : 267.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-[(tert-butoxycarbonyl)amino]-5-methoxybenzoic acid
Inchi Code 1S/C13H17NO5/c1-13(2,3)19-12(17)14-10-6-5-8(18-4)7-9(10)11(15)16/h5-7H,1-4H3,(H,14,17)(H,15,16)
Inchi Key JBXGOBGRVPAQEQ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Grey Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5409

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5409
Availability details for
Quantity
  • Availability
  • Add to Cart