C12H316CEF1A Chemcia Scientific, LLC

2-Amino-6-chloro-N-methoxy-N-methyl-benzamide

  • MDL number MFCD18859461
  • Cas Number : 1339410-44-4
  • Molecular Weight : 214.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-6-chloro-N-methoxy-N-methylbenzamide
Inchi Code 1S/C9H11ClN2O2/c1-12(14-2)9(13)8-6(10)4-3-5-7(8)11/h3-5H,11H2,1-2H3
Inchi Key QJYDXDPTFRJRCP-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin CN
Product Link http://www.chemcia.com/product_fl02.asp?id=5492

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5492
Availability details for
Quantity
  • Availability
  • Add to Cart