C12H316CEF21 Chemcia Scientific, LLC

2-Amino-5-iodo-4-methoxy-benzoic acid

  • MDL number MFCD28968035
  • Cas Number : 1335203-40-1
  • Molecular Weight : 293.06226

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Reddish Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5494

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5494
Availability details for
Quantity
  • Availability
  • Add to Cart