C12H316CEF22 Chemcia Scientific, LLC

5,6-Dichloro-pyridin-2-ylamine

  • MDL number MFCD17012012
  • Cas Number : 1192814-45-1
  • Molecular Weight : 163.00703

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5495

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5495
Availability details for
Quantity
  • Availability
  • Add to Cart