C12H316CEF23 Chemcia Scientific, LLC

4-Cyclopropoxy-benzoic acid

  • MDL number MFCD17000469
  • Cas Number : 62577-90-6
  • Molecular Weight : 178.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(cyclopropyloxy)benzoic acid
Inchi Code 1S/C10H10O3/c11-10(12)7-1-3-8(4-2-7)13-9-5-6-9/h1-4,9H,5-6H2,(H,11,12)
Inchi Key VUANULXCHKBPDD-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5456

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5456
Availability details for
Quantity
  • Availability
  • Add to Cart