C12H316CEF2F Chemcia Scientific, LLC

4-Amino-3-bromo-5-nitro-benzoic acid

  • MDL number MFCD00507339
  • Cas Number : 556651-33-3
  • Molecular Weight : 261.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-amino-3-bromo-5-nitrobenzoic acid
Inchi Code 1S/C7H5BrN2O4/c8-4-1-3(7(11)12)2-5(6(4)9)10(13)14/h1-2H,9H2,(H,11,12)
Inchi Key JCSIZMVUEBODFZ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5488

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5488
Availability details for
Quantity
  • Availability
  • Add to Cart