C12H316CEF33 Chemcia Scientific, LLC

4-Amino-2-chloro-benzaldehyde

  • MDL number MFCD02669926
  • Cas Number : 42460-61-7
  • Molecular Weight : 155.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-amino-2-chlorobenzaldehyde
Inchi Code 1S/C7H6ClNO/c8-7-3-6(9)2-1-5(7)4-10/h1-4H,9H2
Inchi Key PFZVIGWKAMJZJN-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5499

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5499
Availability details for
Quantity
  • Availability
  • Add to Cart