C12H316CEF34 Chemcia Scientific, LLC

2-Methyl-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine

  • MDL number MFCD11858500
  • Cas Number : 100501-59-5
  • Molecular Weight : 137.18582

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5500

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5500
Availability details for
Quantity
  • Availability
  • Add to Cart