C12H316CEF35 Chemcia Scientific, LLC

2-Amino-5-iodo-4-methoxy-benzoic acid methyl ester

  • MDL number MFCD24387259
  • Cas Number : 1256958-34-5
  • Molecular Weight : 307.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-amino-5-iodo-4-methoxybenzoate
Inchi Code 1S/C9H10INO3/c1-13-8-4-7(11)5(3-6(8)10)9(12)14-2/h3-4H,11H2,1-2H3
Inchi Key RXHOMYWGIHJLDD-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5501

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5501
Availability details for
Quantity
  • Availability
  • Add to Cart