C12H316CEF3E Chemcia Scientific, LLC

3-Hydroxy-6-methyl-quinoline-4-carboxylic acid

  • MDL number MFCD21291394
  • Cas Number : 854860-35-8
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-hydroxy-6-methyl-4-quinolinecarboxylic acid
Inchi Code 1S/C11H9NO3/c1-6-2-3-8-7(4-6)10(11(14)15)9(13)5-12-8/h2-5,13H,1H3,(H,14,15)
Inchi Key FYSFKJSKEQONJW-UHFFFAOYSA-N

Description

Purity 90%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5555

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5555
Availability details for
Quantity
  • Availability
  • Add to Cart