C12H316CEF40 Chemcia Scientific, LLC

7-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-benzooxazole-5-carboxylic acid methyl ester

  • MDL number MFCD28968048
  • Molecular Weight : 303.12541

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5557

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5557
Availability details for
Quantity
  • Availability
  • Add to Cart