C12H316CEF41 Chemcia Scientific, LLC

7-Trifluoromethyl-imidazo[1,2-a]pyridine

  • MDL number MFCD11035989
  • Cas Number : 944580-91-0
  • Molecular Weight : 186.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-(trifluoromethyl)imidazo[1,2-a]pyridine
Inchi Code 1S/C8H5F3N2/c9-8(10,11)6-1-3-13-4-2-12-7(13)5-6/h1-5H
Inchi Key TUPNGOKBBWGSGZ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5558

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5558
Availability details for
Quantity
  • Availability
  • Add to Cart