C12H316CEF42 Chemcia Scientific, LLC

7-Trifluoromethyl-imidazo[1,2-a]pyridine-2-carboxylic acid hydrochloride

  • MDL number MFCD28968049
  • Cas Number : 1951439-69-2
  • Molecular Weight : 266.60857

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Salt Data HCl
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5559

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5559
Availability details for
Quantity
  • Availability
  • Add to Cart