C12H316CEF4C Chemcia Scientific, LLC

1H-Pyrazolo[1,5-a]pyrimidin-7-one

  • MDL number MFCD10699578
  • Cas Number : 57489-79-9
  • Molecular Weight : 135.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name pyrazolo[1,5-a]pyrimidin-7(1H)-one
Inchi Code 1S/C6H5N3O/c10-6-2-3-7-5-1-4-8-9(5)6/h1-4,8H
Inchi Key BUEFDJQUUGMUJO-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5561

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5561
Availability details for
Quantity
  • Availability
  • Add to Cart