C12H316CEF51 Chemcia Scientific, LLC

5-Bromo-6-fluoro-1H-indole-2,3-dione

  • MDL number MFCD06409070
  • Cas Number : 118897-99-7
  • Molecular Weight : 244.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-6-fluoro-1H-indole-2,3-dione
Inchi Code 1S/C8H3BrFNO2/c9-4-1-3-6(2-5(4)10)11-8(13)7(3)12/h1-2H,(H,11,12,13)
Inchi Key LNXMNYMNZYJVFX-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5566

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5566
Availability details for
Quantity
  • Availability
  • Add to Cart