C12H316CEF56 Chemcia Scientific, LLC

8-Trifluoromethyl-imidazo[1,2-a]pyridine-2-carboxylic acid ethyl ester

  • MDL number MFCD16987556
  • Cas Number : 1363404-84-5
  • Molecular Weight : 258.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Inchi Code 1S/C11H9F3N2O2/c1-2-18-10(17)8-6-16-5-3-4-7(9(16)15-8)11(12,13)14/h3-6H,2H2,1H3
Inchi Key WZLPRVSGJRYTIQ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5573

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5573
Availability details for
Quantity
  • Availability
  • Add to Cart