C12H316CEF59 Chemcia Scientific, LLC

6,6-Dimethyl-4,5,6,7-tetrahydro-1H-indazole-3-carboxylic acid ethyl ester

  • MDL number MFCD22689252
  • Cas Number : 1423716-54-4
  • Molecular Weight : 222.28946

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5576

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5576
Availability details for
Quantity
  • Availability
  • Add to Cart