C12H316CEF5B Chemcia Scientific, LLC

5,5-Dimethyl-4,5,6,7-tetrahydro-2H-indazole-3-carboxylic acid ethyl ester

  • MDL number MFCD29037500
  • Cas Number : 1887207-39-7
  • Molecular Weight : 222.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 5,5-dimethyl-4,5,6,7-tetrahydro-2H-indazole-3-carboxylate
Inchi Code 1S/C12H18N2O2/c1-4-16-11(15)10-8-7-12(2,3)6-5-9(8)13-14-10/h4-7H2,1-3H3,(H,13,14)
Inchi Key KOEQZIHBLSZFCA-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5578

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5578
Availability details for
Quantity
  • Availability
  • Add to Cart