C12H316CEF5D Chemcia Scientific, LLC

2-Iodo-5-methoxy-4-methyl-phenylamine

  • MDL number MFCD29037501
  • Molecular Weight : 263.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-iodo-5-methoxy-4-methylaniline
Inchi Code 1S/C8H10INO/c1-5-3-6(9)7(10)4-8(5)11-2/h3-4H,10H2,1-2H3
Inchi Key XGYOLHBZQIOBCR-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Red Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5580

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5580
Availability details for
Quantity
  • Availability
  • Add to Cart