C12H316CEF65 Chemcia Scientific, LLC

3-Hydroxy-6-methoxy-quinoline-4-carboxylic acid

  • MDL number MFCD21291405
  • Cas Number : 857758-51-1
  • Molecular Weight : 219.19868

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5599

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5599
Availability details for
Quantity
  • Availability
  • Add to Cart