C12H316CEF67 Chemcia Scientific, LLC

3-Amino-5-bromo-4-hydroxy-benzonitrile

  • MDL number MFCD24644248
  • Cas Number : 862730-24-3
  • Molecular Weight : 213.0347

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5601

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5601
Availability details for
Quantity
  • Availability
  • Add to Cart