C12H316CEF6A Chemcia Scientific, LLC

4-Bromo-2-methyl-benzamide

  • MDL number MFCD12096764
  • Cas Number : 868776-08-3
  • Molecular Weight : 214.06306

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5619

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5619
Availability details for
Quantity
  • Availability
  • Add to Cart