C12H316CEF71 Chemcia Scientific, LLC

2-Chloro-4-fluoro-5-nitro-benzonitrile

  • MDL number MFCD12913468
  • Cas Number : 183325-39-5
  • Molecular Weight : 200.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-4-fluoro-5-nitrobenzonitrile
Inchi Code 1S/C7H2ClFN2O2/c8-5-2-6(9)7(11(12)13)1-4(5)3-10/h1-2H
Inchi Key OUJPFWISNRZAJD-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5627

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5627
Availability details for
Quantity
  • Availability
  • Add to Cart