C12H316CEF72 Chemcia Scientific, LLC

4-Amino-2-chloro-5-nitro-benzonitrile

  • MDL number MFCD26401904
  • Cas Number : 172455-36-6
  • Molecular Weight : 197.58183

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5629

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5629
Availability details for
Quantity
  • Availability
  • Add to Cart