C12H316CEF73 Chemcia Scientific, LLC

(2-Bromo-5-nitro-phenyl)-methanol

  • MDL number MFCD12031825
  • Cas Number : 332883-48-4
  • Molecular Weight : 232.03477

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5638

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5638
Availability details for
Quantity
  • Availability
  • Add to Cart