C12H316CEF76 Chemcia Scientific, LLC

5,5-Dimethyl-4,5,6,7-tetrahydro-1H-indazole-3-carboxylic acid

  • MDL number MFCD20365904
  • Cas Number : 1342547-91-4
  • Molecular Weight : 194.23528

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5644

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5644
Availability details for
Quantity
  • Availability
  • Add to Cart