C12H316CEF77 Chemcia Scientific, LLC

5,5-Dimethyl-4,5,6,7-tetrahydro-2H-indazole-3-carboxylic acid

  • MDL number MFCD22060734
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5,5-dimethyl-4,5,6,7-tetrahydro-2H-indazole-3-carboxylic acid
Inchi Code 1S/C10H14N2O2/c1-10(2)4-3-7-6(5-10)8(9(13)14)12-11-7/h3-5H2,1-2H3,(H,11,12)(H,13,14)
Inchi Key ISMDTJILBJJLPA-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5645

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5645
Availability details for
Quantity
  • Availability
  • Add to Cart