C12H316CEF79 Chemcia Scientific, LLC

6,6-Dimethyl-4,5,6,7-tetrahydro-2H-indazole-3-carboxylic acid

  • MDL number MFCD26935521
  • Cas Number : 1233243-57-6
  • Molecular Weight : 194.23528

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5647

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5647
Availability details for
Quantity
  • Availability
  • Add to Cart