C12H316CEF7A Chemcia Scientific, LLC

3,6-Dibromo-imidazo[1,2-a]pyrazine

  • Cas Number : 1245647-43-1
  • Molecular Weight : 276.91891

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5650

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5650
Availability details for
Quantity
  • Availability
  • Add to Cart