C12H316CEF7B Chemcia Scientific, LLC

6-Bromo-3-chloro-imidazo[1,2-a]pyrazine

  • Cas Number : 1239441-36-1
  • Molecular Weight : 232.46791

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5651

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5651
Availability details for
Quantity
  • Availability
  • Add to Cart