C12H316CEF7C Chemcia Scientific, LLC

5-Methoxy-imidazo[1,2-a]pyridine

  • MDL number MFCD11847485
  • Cas Number : 31409-09-3
  • Molecular Weight : 148.16576

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Grey Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5652

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5652
Availability details for
Quantity
  • Availability
  • Add to Cart