C12H316CEF7F Chemcia Scientific, LLC

6-Hydroxy-nicotinic acid tert-butyl ester

  • MDL number MFCD289680~5C1
  • Cas Number : 1342385-03-8
  • Molecular Weight : 195.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5C
Shipping Temperature Ambient
Country of Origin USA
Product Link http://www.chemcia.com/product_fl02.asp?id=5569

Safety Information

msds http://www.chemcia.com/product_fl02.asp?id=5569
Availability details for
Quantity
  • Availability
  • Add to Cart